Learn More
Medchemexpress LLC Sucrose (standard) | 57-50-1 | 99.8% | 100 MG
SDP
Catalog No. 5000411718
Encompass_Preferred
Sucrose (Standard) is the analytical standard of Sucrose. This product is intended for research and analytical applications. Sucrose (D-(+)-Saccharose) is a disaccharide which is composed of two monosaccharides, glucose and fructose. Sucrose can be applied in some animal models, including metabolic disease, obesity, diet on preference, and diabetes, et al.
  • Grade of analytical standard.
  • Reference standard supplied for assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-11718 Supplier Medchemexpress LLC Supplier No. HYB1779R100MG
Sucrose (Standard) is the analytical standard of Sucrose. This product is intended for research and analytical applications. Sucrose (D-(+)-Saccharose) is a disaccharide which is composed of two monosaccharides, glucose and fructose. Sucrose can be applied in some animal models, including metabolic disease, obesity, diet on preference, and diabetes, et al.
  • Grade of analytical standard.
  • Reference standard supplied for assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC and MS.