Learn More
Sigma Aldrich Fine Chemicals Biosciences Sucrose, 57-50-1, MFCD00006626, 100mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1623637100MG
Encompass
Molecular Formula: C12H22O11, Molecular Weight: 342.30, Format: Neat
Catalog No. NC1393375