Learn More
Perkin Elmer US LLC Sipper Tubing to Cuvette (in), Replacement Tubing
Catalog No. 509042732
Encompass_Preferred
The Sipper Tubing to Cuvette is a teflon hose that connects from a Sipper accessory to the in-flow valve of a Flow-Through cell. With this part number, it can be ordered as a replacement or spare.
Catalog No. 50-904-2732 Supplier Perkin Elmer US LLC Supplier No. B2500129
The Sipper Tubing to Cuvette is a teflon hose that connects from a Sipper accessory to the in-flow valve of a Flow-Through cell. With this part number, it can be ordered as a replacement or spare.