Learn More
Warner Instruments™ Suction Tube, Micro Slit Design
Catalog No. BTX641403 Shop All BTX Harvard Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
BTX641403 Stainless Steel
1 options
Catalog No. BTX641403 Supplier Warner Instruments™ Supplier No. 641403
Micro-slit suction tube is made from 16 gauge Type 316 stainless steel with a 1.65 mm OD and 1.19 mm ID.
  • This tube design has three equally spaced micro-slits at the front entry face and one micro-slit on the rear side of the sipper for air entry. This helps eliminate ground loops and tidal action in the bath and works equally well in all chambers but excels in the larger bath sizes.
Specifications
Type Suction Tube
Material Stainless Steel
Needle Gauge 16 G
Style Micro-slit
Diameter (Metric) 1.65 mm