Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sucunamostat (hydrochloride) | 00-00-0 | 98.1% | 506.89 g/mol | C22H23ClN4O8 | 5 MG
SDP

Catalog No. 5000263259
Encompass_Preferred

Sucunamostat hydrochloride is the hydrochloride salt of sucunamostat, a reversible enteropeptidase inhibitor supplied for research use. It is formulated as a high-purity, small-scale reagent intended for in vitro and preclinical studies.

  • Molecular formula C22H23ClN4O8.
  • Molecular weight 506.89 g/mol.
  • Purity 98.1%.
  • Target enteropeptidase with low-nanomolar potency (IC50 ~4.6-5.4 nM).
  • Chemical form hydrochloride salt for improved solubility.
  • Available in small pack sizes suitable for lab-scale use.

Catalog No. 50-002-63259 Supplier Medchemexpress LLC Supplier No. HY132841A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sucunamostat hydrochloride is the hydrochloride salt of sucunamostat, a reversible enteropeptidase inhibitor supplied for research use. It is formulated as a high-purity, small-scale reagent intended for in vitro and preclinical studies.

  • Molecular formula C22H23ClN4O8.
  • Molecular weight 506.89 g/mol.
  • Purity 98.1%.
  • Target enteropeptidase with low-nanomolar potency (IC50 ~4.6-5.4 nM).
  • Chemical form hydrochloride salt for improved solubility.
  • Available in small pack sizes suitable for lab-scale use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.