Learn More
Medchemexpress LLC Sucunamostat (hydrochloride) | 00-00-0 | 98.1% | C22H23ClN4O8 | 10MG
SDP
Catalog No. 5000259721
Encompass_Preferred
Sucunamostat hydrochloride is an orally active, reversible enteropeptidase inhibitor used in biochemical and pharmacological research to investigate protein digestion and related metabolic pathways. It is supplied as a hydrochloride salt for consistent handling and formulation in laboratory studies.
  • Orally active, reversible enteropeptidase inhibitor.
  • Low-nanomolar potency against rat and human enteropeptidase (reported IC50s).
  • Suitable for in vitro and in vivo studies of protein digestion and metabolic pathways.
  • Provided as a stable hydrochloride salt for reproducible handling.
  • High purity suitable for research applications (reported purity 98.1%).
Catalog No. 50-002-59721 Supplier Medchemexpress LLC Supplier No. HY132841A10MG
Sucunamostat hydrochloride is an orally active, reversible enteropeptidase inhibitor used in biochemical and pharmacological research to investigate protein digestion and related metabolic pathways. It is supplied as a hydrochloride salt for consistent handling and formulation in laboratory studies.
  • Orally active, reversible enteropeptidase inhibitor.
  • Low-nanomolar potency against rat and human enteropeptidase (reported IC50s).
  • Suitable for in vitro and in vivo studies of protein digestion and metabolic pathways.
  • Provided as a stable hydrochloride salt for reproducible handling.
  • High purity suitable for research applications (reported purity 98.1%).