Learn More
Description
Sudan Black B has been used in brain BLAQ (Block Lipids and Aldehyde Quench) technique and is used in lipid detection.
Specifications
Specifications
| Chemical Name or Material | Sudan Black B |
| CAS | 4197-25-5 |
| Color | Black to Black/Brown |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 456.54 g/mol |
| Grade | Certified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.