Learn More
Abcam Sudan black B, Fat-soluble diazo dye, 25G
Catalog No. 502704763
Encompass_Preferred
MW 456.55 g/mol. Fat-soluble diazo dye primarily used for staining of neutral triglycerides and lipids. Sudan dyes are a group of lipid soluble dyes often called lysochromes. Sudan Black B is not as lipid-specific as some of the other Sudan dyes and may be useful to stain other materials. Useful for detecting fats that are contaminated with oil and grease. Used in the study of haematological disorders as it will stain myeloblasts, but not lymphoblasts.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4763 Supplier Abcam Supplier No. AB14628425G
MW 456.55 g/mol. Fat-soluble diazo dye primarily used for staining of neutral triglycerides and lipids. Sudan dyes are a group of lipid soluble dyes often called lysochromes. Sudan Black B is not as lipid-specific as some of the other Sudan dyes and may be useful to stain other materials. Useful for detecting fats that are contaminated with oil and grease. Used in the study of haematological disorders as it will stain myeloblasts, but not lymphoblasts.
The product is subject to the following: Abcam Restricted Use Statement