Learn More
Medchemexpress LLC Sudan I | 842-07-9 | 99.8% | 100 G
SDP
Supplier:  Medchemexpress LLC HYD0024100G
Encompass_Preferred
Sudan I (Solvent Yellow 14) is a diazo-conjugate red dye commonly used as an additive in products such as oils, solvents, and polishes. This monoazo dye, with the chemical formula 1-phenylazo-2-naphthol, is known for inhibiting the growth of bacterial strains *Clostridium perfringens* and *L. rhamnosus*. It is classified as a potential mutagen and carcinogen for humans, capable of inducing genotoxic effects in HepG2 cells and increasing mutagenic and clastogenic effects in MCL-5 cell lines compared to AHH-1 cell lines. It also yields positive results in *Salmonella Typhimurium* mutagenicity tests with S9 activation.
  • Functions as a diazo-conjugate red dye
  • Inhibits *Clostridium perfringens* and *L. rhamnosus* bacterial growth
  • Suitable for use as an additive in oils, solvents, or polishes
  • Exhibits mutagenic and clastogenic effects
  • For research use only
Catalog No. 50-004-10698