Learn More
Medchemexpress LLC Sudan I-d5 (solvent yellow 14-d5) | 752211-63-5 | 99.0% | 253.31 g·mol-1 | C16H7D5N2O | 5 MG
SDP
Catalog No. 5000301839
Encompass_Preferred
Sudan I-d5 is the deuterated isotopologue of Sudan I (Solvent Yellow 14) used as an analytical standard and tracer in quantitative workflows. It is intended for use as an internal standard to improve accuracy and identification in spectrometric and chromatographic analyses.
Catalog No. 50-003-01839 Supplier Medchemexpress LLC Supplier No. HYW0197765MG
Sudan I-d5 is the deuterated isotopologue of Sudan I (Solvent Yellow 14) used as an analytical standard and tracer in quantitative workflows. It is intended for use as an internal standard to improve accuracy and identification in spectrometric and chromatographic analyses.