Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan I 93.0+%, TCI America™
SDP

Catalog No. P058525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P058525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P058525G Supplier TCI America Supplier No. P058525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 842-07-9
Molecular Formula C16H12N2O
Molecular Weight (g/mol) 248.285
MDL Number MFCD00003911
InChI Key ZONYAPYTDIVJGG-VLGSPTGOSA-N
Synonym sudan i, solvent yellow 14, 1-phenylazo-2-naphthol, grasal orange, spirit orange, fast orange, c.i. solvent yellow 14, oil orange, fast oil orange, soudan i
PubChem CID 5858445
IUPAC Name (1Z)-1-(phenylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC=CC=C32

Specifications

Melting Point 129°C
Quantity 25 g
Formula Weight 248.29
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sudan I
EINECSNumber (4)-0985&(5)-3042
RTECSNumber QL4900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.