Learn More
Selleck Chemical LLC Sudan II
SDP
Catalog No. 5000003694
Encompass_Preferred
Sudan II (Sudan Red II Solvent Orange 7 Color Index No 12140 C I 12140) is a lysochrome (fat-soluble dye) azo dye used for staining of triglycerides in frozen sections and some protein bound lipids and lipoproteins on paraffin sections
Catalog No. 50-000-03694 Supplier Selleck Chemical LLC Supplier No. S6240100MG
Add to Cart
Add to Cart
Sudan II (Sudan Red II Solvent Orange 7 Color Index No 12140 C I 12140) is a lysochrome (fat-soluble dye) azo dye used for staining of triglycerides in frozen sections and some protein bound lipids and lipoproteins on paraffin sections