Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan II, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013899 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3899 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3899 Supplier MilliporeSigma Supelco Supplier No. 0793725MG
Add to Cart
Add to Cart

Specifications

CAS 3118-97-6
Linear Formula C18H16N2 O
MDL Number MFCD00003896
Melting Point 156°C to 158°C
Synonym 1-(2,4-Xylylazo)-2-naphthol; Solvent Orange 7
Type Analytical Standard
Quantity 25 mg
Formula Weight 276.33
Percent Purity ≥96% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.