Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sudan III
SCI_ED

Catalog No. S25593 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25593 10 g Solid
S25593A 25 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25593
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 85-86-9
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.397
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Synonym 2-Naphthalenol, Solvent Red 23
PubChem CID 6789251
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43

Specifications

Assay Percent Range >99%
Linear Formula C22H16N4O
Formula Weight 352.4
Packaging Solid
Color Brown to Red
Quantity 10 g
Chemical Name or Material Sudan III
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 4.1, II, : Flammable Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.