Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sudan III-d6 | 1014689-17-8 | 93.0% | 358.43 g/mol | C22H10D6N4O | 10 MG
SDP

Catalog No. 5000297118
Encompass_Preferred

Sudan III-d6 is a deuterium-labeled analogue of the hydrophobic bisazo dye Sudan III, supplied as a research-grade isotopic standard. It contains six deuterium atoms (d6), has formula C22H10D6N4O, and an exact molecular weight of 358.43 g/mol. Typical uses include internal standard and tracer applications in analytical chemistry and mass spectrometry.

  • Deuterium-labeled (d6) isotopic standard.
  • Hydrophobic bisazo dye for analytical use.
  • Purity 93.0% (as provided).
  • Available in small milligram packs for standards and calibration.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-97118 Supplier Medchemexpress LLC Supplier No. HYD0931S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sudan III-d6 is a deuterium-labeled analogue of the hydrophobic bisazo dye Sudan III, supplied as a research-grade isotopic standard. It contains six deuterium atoms (d6), has formula C22H10D6N4O, and an exact molecular weight of 358.43 g/mol. Typical uses include internal standard and tracer applications in analytical chemistry and mass spectrometry.

  • Deuterium-labeled (d6) isotopic standard.
  • Hydrophobic bisazo dye for analytical use.
  • Purity 93.0% (as provided).
  • Available in small milligram packs for standards and calibration.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.