Learn More
Medchemexpress LLC Sudan III-d6 | 1014689-17-8 | 93.0% | 5 MG
Supplier: Medchemexpress LLC HYD0931S5MG
Sudan III-d6 is a deuterium-labeled Sudan III. Sudan III is a hydrophobic bisazo dye. It is intended for research use only and not for sale to patients.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules can be used as tracers for quantitation during drug development.
- Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.