Learn More
Med Vet International Sudan III Fecal Stain, 250ml, J0325
Catalog No. 5000582013
Encompass_Preferred
A quick diagnostic test is to use Sudan III stain as an aid in the assessment of maldigestion/malabsorption syndromes. The Sudan III will selectively stain lipid droplets orange on a fecal smear. 250ml.
Catalog No. 50-005-82013 Supplier Med Vet International Supplier No. JOR325
A quick diagnostic test is to use Sudan III stain as an aid in the assessment of maldigestion/malabsorption syndromes. The Sudan III will selectively stain lipid droplets orange on a fecal smear. 250ml.