Learn More
Electron Microscopy Sciences Sudan Iii Isopropanol Solution 500 ML
SDP
Catalog No. 5031841
Encompass_Preferred
Used in Fat Stains in Supersaturated Alcohol for fat and lipid stain. See Lillie, R.D.,Histopathologic Technical and Practical Histochemistry, 3rd edition, McGraw-Hill, NY, 1965 or Clark, G., (ed) Staining Procedures, 3rd edition, Williams & Wilkins, Baltimore, p. 149, c. 1973
Catalog No. 50-318-41 Supplier Electron Microscopy Sciences Supplier No. 2650303
Used in Fat Stains in Supersaturated Alcohol for fat and lipid stain. See Lillie, R.D.,Histopathologic Technical and Practical Histochemistry, 3rd edition, McGraw-Hill, NY, 1965 or Clark, G., (ed) Staining Procedures, 3rd edition, Williams & Wilkins, Baltimore, p. 149, c. 1973