Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan III, MP Biomedicals™

Catalog No. MP215208725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215208725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215208725 Supplier MP Biomedicals, Inc Supplier No. 0215208725
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in coloring oils, sprit and lacquers. Sudan III, MP Biomedicals is a stain that is also used for zoological, pathological and vegetable objects (wax, cutin, resin, contents of lactiferous ducts, etc.) which are colored red, while cellulose membranes

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-86-9
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.397
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Synonym sudan iii, c.i. solvent red 23, 2-naphthalenol, 1-4-phenylazo phenyl azo, fhninjwbtrxebc-qszpnpogsa-n, sudaniii, sudan iiisolvent red 23, 2-naphthalenol, 1-2-4-2-phenyldiazenyl phenyl diazenyl, 1-2-4-2-phenyldiazen-1-yl phenyl diazen-1-yl naphthalen-2-ol, 1-e-4-e-phenylazo phenyl azo naphthalen-2-ol, 1-4-phenyldiazenylphenyl hydrazinylidene naphthalen-2-one
PubChem CID 6789251
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43

Specifications

Melting Point 199°C (literature)
Quantity 25 g
Solubility Information Insoluble in water; soluble in chloroform, glacial acetic acid; moderately soluble in alcohol, ether, acetone, fixed and volatile oils, hot glycerol. Clear red solution at 1mg/mL in toluence.
Formula Weight 352.4
Physical Form Powder
Recommended Storage Room Temperature (15–30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.