Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan III, MP Biomedicals™

Catalog No. ICN15208780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15208780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15208780 Supplier MP Biomedicals, Inc Supplier No. 0215208780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lipophilic dye used as a biological stain for histological applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-86-9
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.397
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Synonym 1-[4-(phenylazo)phenylazo]-2-naphthol, Solvent Red 23
PubChem CID 6789251
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43

Specifications

Color Brown/Red
Quantity 100 g
Physical Form Powder
Formula Weight 352.4
Grade Not Available
Chemical Name or Material Sudan III
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.