Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan III, Saturated, Vion Biosciences
SCI_ED

Catalog No. S02485 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02485 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02485 Supplier Aldon Corporation Supplier No. VNSS1050100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Effectively visualize and identify lipid in tissue specimens with the red-orange histological stain: saturated Sudan III in IPA.

  • Effectively visualize and identify lipid in tissue specimens with the red-orange histological stain: saturated Sudan III in IPA.

Chemical Identifiers

CAS 85-86-9 , 67-63-0
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.40
MDL Number MFCD00003905
InChI Key HTPQPMPFXUWUOT-SRURNFRUSA-N
Synonym Chromatography Dye Solution - Red
PubChem CID Data not available.
IUPAC Name (1Z)-1-{2-[4-(2-phenyldiazen-1-yl)phenyl]hydrazin-1-ylidene}-1,2-dihydronaphthalen-2-one
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point -90°C
Boiling Point ~ 82°C
CAS Min % 1%, 99%
Concentration 1.00%
Dye Content 1.00%
Identification 1% Sudan III in IPA
CAS Max % 99.00%, 1%
Infrared Spectrum Yes
Odor Aromatic odor.
Packaging HDPE Bottle
Specific Gravity 0.786 to 0.789 (Isopropyl Alcohol)
Assay Percent Range 99.00%
Vapor Pressure 33 mm (at 20°C)
Certificate of Analysis Yes
UN Number UN1219
Decomposition Information Data not available.
Density 0.786 to 0.789 (Isopropyl Alcohol)
pH Color Changes Data not available.
Solvent or Matrix Isopropanol
Color Orange/Red
Solubility Information Complete in water.
pH Data not available.
Quantity 100 mL
Flash Point 12°C
Physical Form Data not available.
Percent Purity 1.00%
Grade Data not available.
Concentration or Composition (by Analyte or Components) 1.00%
Chemical Name or Material Sudan III
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Causes serious eye irritation.
Hazard Statement May cause drowsiness or dizziness.
ShelfLife 3 years
DOTInformation UN1219, Isopropanol solution, 3(II), Limited quantity equal to or less than 1 L
EINECSNumber 201-638-4
RTECSNumber QK4250000
Recommended Storage Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.