Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan IV, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015694 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5694 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5694 Supplier MilliporeSigma Supelco Supplier No. 6738625MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 85-83-6
Physical Form Neat
Linear Formula C24H20N4 O
MDL Number MFCD00003893
Melting Point 199°C
Synonym Biebrich scarlet R fat soluble; Fat Ponceau R or 4; Lipid Crimson; Oil Red IV; Scarlet Red Scharlach; Solvent Red 24; Sudan R or BB
Type Analytical Standard
Quantity 25 mg
Formula Weight 380.44
Percent Purity ≥96% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.