Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Sudan IV, Saturated, Vion Biosciences
SCI_ED

Catalog No. S02484 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02484 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02484 Supplier Innovating Science™ Supplier No. VNSS1070100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Effectively visualize and identify lipid in tissue specimens with the deep red histological stain: Saturated Sudan IV in IPA.

  • Effectively visualize and identify lipid in tissue specimens with the deep red histological stain: Saturated Sudan IV in IPA.

Chemical Identifiers

CAS 85-83-5 , 67-63-0
Molecular Formula Mixture
Molecular Weight (g/mol) 380.44
MDL Number MFCD00003893
InChI Key KMDLOETUWUPGMB-BXCCFQQFSA-N
Synonym Chromatography Dye Solution - Red
PubChem CID Data not available.
ChEBI CHEBI:88014
IUPAC Name (1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronapthalen-2-one
SMILES CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C

Specifications

Melting Point -50°C
Boiling Point 85°C to 100°C
CAS Min % 0.5%, 99.5%
CAS Max % 99.5%, 0.5%
Concentration 0.50%
Dye Content 0.50%
Assay Percent Range 99.50%
Extinction Coefficient 5.7 mM-1 cm-1
UN Number UN1219
Solubility Information Complete in water.
Identification 0.5% Sudan IV in IPA
Infrared Spectrum Yes
Percent Purity 0.50%
Grade Data not available.
Concentration or Composition (by Analyte or Components) 0.50%
Molar Absorption Coefficient 32,500 (at 518 nm)
Odor Aromatic odor.
Packaging HDPE Bottle
Specific Gravity 0.8 (Isopropyl Alcohol)
Vapor Pressure 33 mm (at 20°C)
Certificate of Analysis Yes
Decomposition Information Data not available.
Density 0.8 (Isopropyl Alcohol)
pH Color Changes Data not available.
Solvent or Matrix Isopropanol
Color Orange/Red
pH Data not available.
Quantity 100 mL
Flash Point 12°C
Physical Form Data not available.
Chemical Name or Material Sudan IV
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Causes serious eye irritation.
Hazard Statement May cause drowsiness or dizziness.
ShelfLife 3 years
DOTInformation UN1219, Isopropanol solution, 3(II), Limited quantity equal to or less than 1 L
EINECSNumber 201-635-8
RTECSNumber QL5775000
Recommended Storage Store in a cool, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.