Learn More
G-Biosciences Sudan IV Solution

Description
It is commonly used in histology and cytology to visualize lipids in tissue sections. The dye is lysochrome, which is fat-soluble and binds to lipids, causing them to appear red.
Specifications
Specifications
| CAS | 85-83-6 |
| Color | Red |
| Quantity | 250 mL |
| Molecular Weight (g/mol) | 380.44 g/mol |
| Grade | Research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.