Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan Orange G, MP Biomedicals™

Catalog No. ICN15670425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15670425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15670425 Supplier MP Biomedicals, Inc Supplier No. 0215670425

missing translation for 'validMainframeMsg'

C.I. 11920, (Solvent Orange 1; 4-[Phenylazo]-resorcinol), Dye content: Approx. 95%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-85-6
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002275
InChI Key PULNESMFDLBKAZ-SDNWHVSQSA-N
PubChem CID 5921655
IUPAC Name (4E)-3-hydroxy-4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1

Specifications

Quantity 25 g
Physical Form Solid
Formula Weight 214.2
Chemical Name or Material Sudan Orange G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.