Learn More
Abcam Sudan red 7 B, Diazo dye, 10G
Catalog No. 502704739
Encompass_Preferred
MW 379.47 g/mol. Lipid soluble, diazo dye for staining fats in plant and animal tissues. Member of the Sudan family of hydrophobic lipid-staining dyes. Sudan dyes are a group of lipid soluble dyes often called lysochromes. With the use of certain solvents, may also be used to stain some protein bound lipids in paraffin sections. Has also been used to stain suberin lamellae in plant tissues.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4739 Supplier Abcam Supplier No. AB14630610G
MW 379.47 g/mol. Lipid soluble, diazo dye for staining fats in plant and animal tissues. Member of the Sudan family of hydrophobic lipid-staining dyes. Sudan dyes are a group of lipid soluble dyes often called lysochromes. With the use of certain solvents, may also be used to stain some protein bound lipids in paraffin sections. Has also been used to stain suberin lamellae in plant tissues.
The product is subject to the following: Abcam Restricted Use Statement