Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sudan red 7B | 6368-72-5 | 98.4% | C24H21N5 | 5 G
SDP

Catalog No. 5000353345
Encompass_Preferred

Sudan red 7B is a red non-fluorescent stain primarily used for staining fat bodies. It is stable under recommended storage conditions and suitable for laboratory chemical applications and substance manufacturing.

  • Red non-fluorescent stain
  • Used for staining fat bodies
  • Solid appearance
  • Brown to reddish brown color
  • Molecular weight of 379.46
  • Purity of 98.4%
  • Stable under recommended storage conditions
  • Suitable for laboratory chemical applications and substance manufacturing

Catalog No. 50-003-53345 Supplier Medchemexpress LLC Supplier No. HY1456125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sudan red 7B is a red non-fluorescent stain primarily used for staining fat bodies. It is stable under recommended storage conditions and suitable for laboratory chemical applications and substance manufacturing.

  • Red non-fluorescent stain
  • Used for staining fat bodies
  • Solid appearance
  • Brown to reddish brown color
  • Molecular weight of 379.46
  • Purity of 98.4%
  • Stable under recommended storage conditions
  • Suitable for laboratory chemical applications and substance manufacturing

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.