Learn More
Medchemexpress LLC Sudan red 7B | 6368-72-5 | MFCD00003904 | 98.4% | C24H21N5 | 1 G
SDP
Catalog No. 5000354176
Encompass_Preferred
Sudan red 7B is a red non-fluorescent stain that can be used to stain fat bodies. It appears as a solid with a brown to reddish-brown color.
  • Red non-fluorescent stain
  • Used to stain fat bodies
  • Appears as a solid
  • Brown to reddish-brown color
Catalog No. 50-003-54176 Supplier Medchemexpress LLC Supplier No. HY1456121G
Sudan red 7B is a red non-fluorescent stain that can be used to stain fat bodies. It appears as a solid with a brown to reddish-brown color.
  • Red non-fluorescent stain
  • Used to stain fat bodies
  • Appears as a solid
  • Brown to reddish-brown color