Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan Red 7B, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015005 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5005 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5005 Supplier MilliporeSigma Supelco Supplier No. 5337325MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 6368-72-5
Physical Form Neat
Linear Formula C24H21N5
MDL Number MFCD00003904
Melting Point 130°C
Synonym Fat Red 7 B; Fat red bluish; Solvent Red 19
Type Analytical Standard
Quantity 25 mg
Formula Weight 379.46
Percent Purity ≥96% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.