Learn More
ATCC SU-DHL-1; Histiocytic Lymphoma; Human (Homo sapiens)
Supplier:  ATCC CRL2955
Encompass
SU-DHL-1 is a histiocytic cell that was isolated from the lymph node of a White, 10-year-old, male patient with large cell lymphoma. This cell line was deposited by A Epstein and can be used in immunology research.
Catalog No. 50-238-2246