Learn More
ATCC SU-DHL-16; B Cell Lymphoma; Human (Homo sapiens)
Catalog No. 502383193
Encompass
SU-DHL-16 is a B lymphocyte cell line that was isolated in 1980 from the lymph node of a male with large cell lymphoma. It has applications in immunology research.
Catalog No. 50-238-3193 Supplier ATCC Supplier No. CRL2964
SU-DHL-16 is a B lymphocyte cell line that was isolated in 1980 from the lymph node of a male with large cell lymphoma. It has applications in immunology research.