Learn More
ATCC SU-DHL-2; B Cell Lymphoma; Human (Homo sapiens)
Catalog No. 502384654
Encompass
SU-DHL-2 is a histiocytic cell line that was isolated in 1974 from the lymph node of a White 73-year-old female with large cell lymphoma. This cell line can be used in immunology research.
Catalog No. 50-238-4654 Supplier ATCC Supplier No. CRL2956
SU-DHL-2 is a histiocytic cell line that was isolated in 1974 from the lymph node of a White 73-year-old female with large cell lymphoma. This cell line can be used in immunology research.