Learn More
ATCC SU-DHL-4; Large Cell Lymphoma; Human (Homo sapiens)
Catalog No. 502382251
Encompass
SU-DHL-4 is a lymphoblast-like cell that was isolated from the peritoneal effusion of a White, 38-year-old, male patient. This cell line was deposited by A Epstein in 1976 and can be used in immunology research.
Catalog No. 50-238-2251 Supplier ATCC Supplier No. CRL2957
SU-DHL-4 is a lymphoblast-like cell that was isolated from the peritoneal effusion of a White, 38-year-old, male patient. This cell line was deposited by A Epstein in 1976 and can be used in immunology research.