Learn More
Cayman Chemical Sudan Red 7B, CAS: 6368-72-5, 10 g
SDP
Catalog No. 502947160
Encompass_Preferred
An azo dye; abs = 525 nm; has been used to stain lipids in hepatocytes and suberin lamellae in plant cell walls; has been included as a standard in LC-MS/MS-based protocols for the detection of banned colorants in foodstuffs
Catalog No. 50-294-7160 Supplier Cayman Chemical Supplier No. 3331510 G
An azo dye; abs = 525 nm; has been used to stain lipids in hepatocytes and suberin lamellae in plant cell walls; has been included as a standard in LC-MS/MS-based protocols for the detection of banned colorants in foodstuffs