Learn More
Cayman Chemical Sudn Red 7B 50g
SDP
Catalog No. 502948256
Encompass_Preferred
An azo dye; abs = 525 nm; has been used to stain lipids in hepatocytes and suberin lamellae in plant cell walls; has been included as a standard in LC-MS/MS-based protocols for the detection of banned colorants in foodstuffs
Catalog No. 50-294-8256 Supplier Cayman Chemical Supplier No. 3331550 G
An azo dye; abs = 525 nm; has been used to stain lipids in hepatocytes and suberin lamellae in plant cell walls; has been included as a standard in LC-MS/MS-based protocols for the detection of banned colorants in foodstuffs