Learn More
TARGETMOL CHEMICALS INC SUDOXICAM 5MG
SDP
Catalog No. 502581135
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sudoxicam (4-Hydroxy-2-methyl-N-2-thiazolyl-2H-12-benzothiazine-3-carboxamide 11-dioxide) is a reversible antagonist of COX with anti-inflammatory anti-edema and antipyretic properties. purity: 100%
Catalog No. 50-258-1135 Supplier TARGETMOL CHEMICALS INC Supplier No. T169505MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sudoxicam (4-Hydroxy-2-methyl-N-2-thiazolyl-2H-12-benzothiazine-3-carboxamide 11-dioxide) is a reversible antagonist of COX with anti-inflammatory anti-edema and antipyretic properties. purity: 100%