Learn More
Sterlitech Corporation SUEZ (GE) Flat Sheet Membrane, DK, PA-TFC, NF, 305 x 305mm, 1/Pk
SDP
Supplier:  Sterlitech Corporation YMDKSP3001
Encompass_Preferred
The DK designation is commonly used for Acid Purification, Alcohol Purification, Antibiotic Concentration, BOD/COD Reduction, Cheese Whey Desalting, Detergent Removal, Dextrose Purification, Dye Concentration, Ethylene Glycol Purification, Heavy Metal Removal, Plating Waste, Polysaccharide Desalting, Sugar Fractionation.
Catalog No. 50-195-2337