Learn More
Aviva Systems Biology Corp SUFU Antibody - middle region (ARP39233_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39233_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUFU. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SUFU. Alternate names: PRO1280, SUFUH, SUFUXLApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56953