Learn More
Medchemexpress LLC HY-100209 5mg Medchemexpress, Sufugolix CAS:308831-61-0 Purity:>98%
SDP
Catalog No. 501931751
Encompass_Preferred
Medchemexpress, HY-100209 5mg Sufugolix CAS:308831-61-0 Sufugolix (TAK-013) is a highly potent and orally available luteinizing hormone-releasing hormone ( LHRH ) receptor antagonist with an IC 50 of 0.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1751 Supplier Medchemexpress LLC Supplier No. HY1002095MG
Medchemexpress, HY-100209 5mg Sufugolix CAS:308831-61-0 Sufugolix (TAK-013) is a highly potent and orally available luteinizing hormone-releasing hormone ( LHRH ) receptor antagonist with an IC 50 of 0.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.