Learn More
Medchemexpress LLC HY-B0079 10mg Medchemexpress, Sugammadex (sodium) CAS:343306-79-6 Purity:>98%
SDP
Catalog No. 502082355
Encompass_Preferred
Medchemexpress, HY-B0079 10mg Sugammadex (sodium) CAS:343306-79-6 Sugammadex sodium is a synthetic γ-cyclodextrin derivative, and acts as a new reversal agent for neuromuscular block. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2355 Supplier Medchemexpress LLC Supplier No. HYB007910MG
Medchemexpress, HY-B0079 10mg Sugammadex (sodium) CAS:343306-79-6 Sugammadex sodium is a synthetic γ-cyclodextrin derivative, and acts as a new reversal agent for neuromuscular block. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.