Learn More
TARGETMOL CHEMICALS INC SUGAMMADEX SODIUM 50MG
SDP
Catalog No. 502576693
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Sugammadex sodium (Org25969) a synthetic derivative of (gamma)-cyclodextrin is a steroid-based neuromuscular blocker reversing agent. purity: 99%
Catalog No. 50-257-6693 Supplier TARGETMOL CHEMICALS INC Supplier No. T532650MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Sugammadex sodium (Org25969) a synthetic derivative of (gamma)-cyclodextrin is a steroid-based neuromuscular blocker reversing agent. purity: 99%