Learn More
NEOGEN Sugar Free Agar, 500 g
SDP
Catalog No. 502015891
Encompass_Preferred
Sugar Free Agar is used for the enumeration of psychrotrophic and mesophilic Gram-negative rods, and is not intended for use in the diagnosis of disease or other conditions in humans.A formula described by the International Dairy Federation for the enumeration of psychrotrophic and mesophilic Gram negative rods in butter and other dairy products. The Gram negative rods are able to deaminate proteins as a carbon source, whilst some enterococci are inhibited by this formula. The medium conforms to the formulation of the International Dairy Federation (I.D.F.).Formula:Gelatin Peptone - 7.5 g/LTryptone - 7.5 g/LSodium Chloride - 5.0 g/LAgar - 14.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/sugar-free-agar
Catalog No. 50-201-5891 Supplier NEOGEN Supplier No. 700004764
Sugar Free Agar is used for the enumeration of psychrotrophic and mesophilic Gram-negative rods, and is not intended for use in the diagnosis of disease or other conditions in humans.A formula described by the International Dairy Federation for the enumeration of psychrotrophic and mesophilic Gram negative rods in butter and other dairy products. The Gram negative rods are able to deaminate proteins as a carbon source, whilst some enterococci are inhibited by this formula. The medium conforms to the formulation of the International Dairy Federation (I.D.F.).Formula:Gelatin Peptone - 7.5 g/LTryptone - 7.5 g/LSodium Chloride - 5.0 g/LAgar - 14.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/sugar-free-agar