Learn More
Waters Corp Waters Sugar-Pak Guard-Pak Insert, 10 μm, HPLC, L19
Catalog No. 50785075 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Guard-Pak Insert is designed to extend the lifetime of analytical columns by removing chemical and particulate contaminants from the mobile phase, thereby enhancing equipment performance. It is suitable for 10 μm particle size columns.
  • Utilizes sorbents identical to HPLC columns for optimal compatibility.
  • Contains microparticulate cation-exchange gel in calcium form.
  • Manufactured under stringent conditions with strict QC for reproducibility.
  • Each batch chromatographically tested; individually quality tested.
  • Tolerates pressures up to 6,000 psi (415 bar); optimal at 4,000 to 5,000 psi.
  • Requires miscible eluents to prevent precipitation.
  • Features spherical particle shape and low silanol activity for HPLC systems.
  • USP Classification L19, ideal for carbohydrate applications.
Catalog No. 50-785-075 Supplier Waters Corp Supplier No. WAT015209
This Guard-Pak Insert is designed to extend the lifetime of analytical columns by removing chemical and particulate contaminants from the mobile phase, thereby enhancing equipment performance. It is suitable for 10 μm particle size columns.
  • Utilizes sorbents identical to HPLC columns for optimal compatibility.
  • Contains microparticulate cation-exchange gel in calcium form.
  • Manufactured under stringent conditions with strict QC for reproducibility.
  • Each batch chromatographically tested; individually quality tested.
  • Tolerates pressures up to 6,000 psi (415 bar); optimal at 4,000 to 5,000 psi.
  • Requires miscible eluents to prevent precipitation.
  • Features spherical particle shape and low silanol activity for HPLC systems.
  • USP Classification L19, ideal for carbohydrate applications.