Learn More
Waters Corp Sugar-Pak I Column, 10 µm, 6.5 mm X 300 mm, 1/pk
Catalog No. 50818699 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sugar Pak I column separates monosaccharides and sugar alcohols via a strong cation-exchange mechanism. The resin is based on a sulfonated styrene-divinylbenzene polymer that provides pH stability by means of a calcium counterion.
Catalog No. 50-818-699 Supplier Waters Corp Supplier No. WAT085188
The Sugar Pak I column separates monosaccharides and sugar alcohols via a strong cation-exchange mechanism. The resin is based on a sulfonated styrene-divinylbenzene polymer that provides pH stability by means of a calcium counterion.