Learn More
Showa Denko America Inc SUGAR SH1011
Supplier:  Showa Denko America Inc F6378100
Encompass_Preferred
Multimode styrene divinylbenzene based columns with hydrogen counterion, used for simultaneous analysis of saccharides and organic acids, separates neutral sugars in size exclusion mode and organic acids in ion exclusion mode, suitable for the analysis of uronic and aldonic acids, fulfills USP L17 and L22 requirements, 8.0 mm ID x 300 mm L
Catalog No. 50-161-3253