Learn More
Aviva Systems Biology Corp Sugp1 Antibody - N-terminal region (ARP40962_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sugp1. It was validated on Western Blot. . Alternate names: 5730496N02Rik, Sf4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-284-905