Learn More
Aviva Systems Biology Corp SUGT1 Antibody - middle region (ARP52240_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52240_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUGT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SUGT1. Alternate names: SGT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-668