Learn More
Premier Lab Supply 40mm XRF Sample Cups – PN# SC-3340-T
SDP
Catalog No. NC0753588
Encompass_Preferred
40mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-3340-T) – Suitable for Bruker SRS, S4, S8; Thermo Scientific™ ARL™ Perform’X – OD: 40mm; ID: 34mm; Height: 30mm; Volume: 18mL – 100 per bag
Catalog No. NC0753588 Supplier Premier Lab Supply Supplier No. SC3340T
40mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-3340-T) – Suitable for Bruker SRS, S4, S8; Thermo Scientific™ ARL™ Perform’X – OD: 40mm; ID: 34mm; Height: 30mm; Volume: 18mL – 100 per bag