Learn More
Benchmark Products Inc Suite Pad Mop, Non-Particulating, Snag-Resistant, Thick Absorbent Pad, 25 Mops Per Case
SDP
Catalog No. NC9988986
Encompass
The Suite Pad Mop is a high-performance cleaning solution crafted from synthetic ultra-fibers, designed specifically for critical environments. Its unique, snag-resistant, and non-particulating structure ensures exceptional contamination removal, achieving over 98% surface cleanliness. Ideal for walls, ceilings, and floors, this non-sterile mop pad excels in ISO Class 5 cleanrooms, providing reliable durability and effective cleaning capabilities that distinguish it from conventional mop pads.
Catalog No. NC9988986 Supplier Benchmark Products Inc Supplier No. YGLPP51525
The Suite Pad Mop is a high-performance cleaning solution crafted from synthetic ultra-fibers, designed specifically for critical environments. Its unique, snag-resistant, and non-particulating structure ensures exceptional contamination removal, achieving over 98% surface cleanliness. Ideal for walls, ceilings, and floors, this non-sterile mop pad excels in ISO Class 5 cleanrooms, providing reliable durability and effective cleaning capabilities that distinguish it from conventional mop pads.