Learn More
Benchmark Products Inc Suite Primary Duty Mop Pad, Engineered Fiber, Effective Contaminant Removal, 25 mop pads
SDP
Catalog No. NC1399213
Encompass
The Suite Primary Duty Mop Pad is engineered for efficient surface cleaning and disinfectant application, designed specifically for the Suite Touchless Primary Duty Mop System. Its non-sterile, engineered fiber composition enhances cleaning efficiency by ensuring liberal distribution of cleaning agents. This product minimizes cross-contamination and reduces operator handling, leading to significantly decreased pad changeover time and improved operational efficiency in controlled environments.
This item is not returnable. View return policy
Catalog No. NC1399213 Supplier Benchmark Products Inc Supplier No. YTL51725
The Suite Primary Duty Mop Pad is engineered for efficient surface cleaning and disinfectant application, designed specifically for the Suite Touchless Primary Duty Mop System. Its non-sterile, engineered fiber composition enhances cleaning efficiency by ensuring liberal distribution of cleaning agents. This product minimizes cross-contamination and reduces operator handling, leading to significantly decreased pad changeover time and improved operational efficiency in controlled environments.